CH4309656787 ChemBridge Corporation

8-fluoro-4-oxo-1,4-dihydro-2-quinolinecarboxylic acid

  • MDL number MFCD09929424
  • Cas Number : 1016506-97-0
  • Molecular Weight : 207.1579

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040067

Safety Information

MSDS https://www.hit2lead.com/sds/4040067_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart