CH4309656798 ChemBridge Corporation

2-methoxy-4,5-dimethylbenzenesulfonyl chloride

  • MDL number MFCD08443668
  • Cas Number : 90416-52-7
  • Molecular Weight : 234.6998

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040135

Safety Information

signalword Danger
MSDS https://www.hit2lead.com/building-blocks/4040135
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart