CH4309656803 ChemBridge Corporation

5-methyl-1-propyl-1H-1,2,4-triazol-3-amine

  • MDL number MFCD16556129
  • Cas Number : 1227465-52-2
  • Molecular Weight : 140.1863

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040176

Safety Information

MSDS https://www.hit2lead.com/sds/4040176_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart