CH4309656812 ChemBridge Corporation

5-(5-amino-1,3,4-thiadiazol-2-yl)-2-pyrrolidinone

  • MDL number MFCD18381916
  • Cas Number : 1256643-02-3
  • Molecular Weight : 184.2189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040219

Safety Information

MSDS https://www.hit2lead.com/sds/4040219_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart