CH4309656828 ChemBridge Corporation

5-cyclopropyl-1,2,4-oxadiazol-3-amine

  • MDL number MFCD11847813
  • Cas Number : 868696-42-8
  • Molecular Weight : 125.1286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040300

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4040300
Availability details for
Quantity
  • Availability
  • Add to Cart