CH4309656831 ChemBridge Corporation

3-(3-bromo-1H-1,2,4-triazol-5-yl)propanoic acid

  • MDL number MFCD16556134
  • Cas Number : 933690-20-1
  • Molecular Weight : 220.024

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040328

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4040328
Availability details for
Quantity
  • Availability
  • Add to Cart