CH4309656832 ChemBridge Corporation

5-methyl-3-isoxazolecarbonitrile

  • MDL number MFCD09064940
  • Cas Number : 57351-99-2
  • Molecular Weight : 108.0981

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040331

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4040331_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart