CH4309656834 ChemBridge Corporation

3-phenyl-2,4-imidazolidinedione

  • MDL number MFCD00209486
  • Cas Number : 2221-13-8
  • Molecular Weight : 176.172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040353

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4040353
Availability details for
Quantity
  • Availability
  • Add to Cart