CH4309656846 ChemBridge Corporation

phenyl(4-piperidinyl)methanol

  • MDL number MFCD05864581
  • Cas Number : 38081-60-6
  • Molecular Weight : 191.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4005907

Safety Information

MSDS https://www.hit2lead.com/sds/4005907_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart