CH4309656864 ChemBridge Corporation

5-(1H-tetrazol-1-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD12028417
  • Cas Number : 1039008-40-6
  • Molecular Weight : 180.1243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4006024

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4006024
Availability details for
Quantity
  • Availability
  • Add to Cart