CH4309656890 ChemBridge Corporation

7-chloro-1,3-benzoxazol-2-amine

  • MDL number MFCD01664219
  • Cas Number : 64037-11-2
  • Molecular Weight : 168.5804

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040807

Safety Information

MSDS https://www.hit2lead.com/sds/4040807_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart