CH4309656898 ChemBridge Corporation

3-bromo-6-methoxypyridazine

  • MDL number MFCD09702456
  • Cas Number : 17321-29-8
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040879

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4040879
Availability details for
Quantity
  • Availability
  • Add to Cart