CH4309656970 ChemBridge Corporation

1-benzyl-1H-indole-4-carbonitrile

  • MDL number MFCD15200968
  • Cas Number : 177548-00-4
  • Molecular Weight : 232.2799

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041332

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4041332_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart