CH4309656990 ChemBridge Corporation

1-oxo-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid

  • MDL number MFCD00483631
  • Cas Number : 63586-82-3
  • Molecular Weight : 191.1834

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041444

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4041444
Availability details for
Quantity
  • Availability
  • Add to Cart