CH4309657035 ChemBridge Corporation

3-(1H-1,2,4-triazol-1-ylmethyl)aniline

  • MDL number MFCD08060528
  • Cas Number : 127988-22-1
  • Molecular Weight : 174.2025

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4011704

Safety Information

MSDS https://www.hit2lead.com/sds/4011704_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart