CH4309657037 ChemBridge Corporation

(1Z)-N'-hydroxy-2-(methylthio)ethanimidamide

  • MDL number MFCD09842259
  • Cas Number : 104608-67-5
  • Molecular Weight : 120.1734

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042689

Safety Information

MSDS https://www.hit2lead.com/sds/4042689_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart