CH4309657069 ChemBridge Corporation

6,7-dihydro-5H-cyclopenta[d]pyrimidin-4-amine

  • MDL number MFCD00603725
  • Cas Number : 26979-06-6
  • Molecular Weight : 135.1665

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013921

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4013921
Availability details for
Quantity
  • Availability
  • Add to Cart