CH4309657075 ChemBridge Corporation

1-aminocycloheptanecarboxamide

  • MDL number MFCD10686671
  • Cas Number : 906075-26-1
  • Molecular Weight : 156.2254

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043046

Safety Information

MSDS https://www.hit2lead.com/sds/4043046_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart