CH4309657162 ChemBridge Corporation

5-piperidin-1-yl-2-furaldehyde

  • MDL number MFCD02614962
  • Cas Number : 22868-60-6
  • Molecular Weight : 179.2157

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4300151

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4300151
Availability details for
Quantity
  • Availability
  • Add to Cart