CH4309657177 ChemBridge Corporation

(pyridin-2-ylthio)acetic acid

  • MDL number MFCD00276114
  • Cas Number : 10002-29-6
  • Molecular Weight : 169.201

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4400385

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4400385
Availability details for
Quantity
  • Availability
  • Add to Cart