CH4309657183 ChemBridge Corporation

2-oxo-2,5,6,7-tetrahydro-1H-cyclopenta[b]pyridine-3-carboxylic acid

  • MDL number MFCD07801152
  • Cas Number : 115122-63-9
  • Molecular Weight : 179.1727

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4401889

Safety Information

MSDS https://www.hit2lead.com/sds/4401889_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart