CH4312834595 ChemBridge Corporation

(2,5-dimethyl-3-furyl)methanol

  • MDL number MFCD11646256
  • Cas Number : 1003-96-9
  • Molecular Weight : 126.1531

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034575

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4034575
Availability details for
Quantity
  • Availability
  • Add to Cart