CH4312834603 ChemBridge Corporation

tetrahydro-3-thiophenecarbonitrile 1,1-dioxide

  • MDL number MFCD06809749
  • Cas Number : 17389-09-2
  • Molecular Weight : 145.1796

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035545

Safety Information

MSDS https://www.hit2lead.com/sds/4035545_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart