CH4312834605 ChemBridge Corporation

2-hydrazino-4-methoxypyrimidine

  • MDL number MFCD02734394
  • Cas Number : 89181-80-6
  • Molecular Weight : 140.1432

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035787

Safety Information

MSDS https://www.hit2lead.com/sds/4035787_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart