CH4312834630 ChemBridge Corporation

tert-butyl 2,7-diazaspiro[4.5]decane-2-carboxylate

  • MDL number MFCD08234437
  • Cas Number : 885268-42-8
  • Molecular Weight : 240.3419

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4037160

Safety Information

MSDS https://www.hit2lead.com/sds/4037160_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart