CH4312834641 ChemBridge Corporation

2-methyl-2,7-diazaspiro[4.5]decane

  • MDL number MFCD11053747
  • Cas Number : 1086395-66-5
  • Molecular Weight : 154.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4038747

Safety Information

MSDS https://www.hit2lead.com/sds/4038747_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart