CH4312834654 ChemBridge Corporation

3-chloro-1H-indole-2-carboxylic acid

  • MDL number MFCD11007738
  • Cas Number : 28737-32-8
  • Molecular Weight : 195.6024

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042558

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4042558
Availability details for
Quantity
  • Availability
  • Add to Cart