CH4312834660 ChemBridge Corporation

2-methyl-5-[5-(trifluoromethyl)-3-isoxazolyl]benzenesulfonyl chloride

  • MDL number MFCD09743338
  • Cas Number : 936074-62-3
  • Molecular Weight : 325.6914

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042607

Safety Information

signalword Danger
hcodes 8
MSDS https://www.hit2lead.com/sds/4042607_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart