CH4312834666 ChemBridge Corporation

2-chloro-3-(1,2,4-oxadiazol-3-yl)pyridine

  • MDL number MFCD18447664
  • Cas Number : 1268334-78-6
  • Molecular Weight : 181.5792

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042679

Safety Information

MSDS https://www.hit2lead.com/sds/4042679_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart