CH4312834680 ChemBridge Corporation

N,N-dimethyl-1,3,5-triazine-2,4-diamine

  • MDL number MFCD00085979
  • Cas Number : 4039-98-9
  • Molecular Weight : 139.1585

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042889

Safety Information

MSDS https://www.hit2lead.com/sds/4042889_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart