CH4312834693 ChemBridge Corporation

2-methyl-5-[5-(trifluoromethyl)-3-isoxazolyl]-3-thiophenesulfonyl chloride

  • MDL number MFCD15729946
  • Cas Number : 1268334-82-2
  • Molecular Weight : 331.7191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043094

Safety Information

signalword Danger
hcodes 8
MSDS https://www.hit2lead.com/sds/4043094_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart