CH4312834724 ChemBridge Corporation

3-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-2-fluoropyridine

  • MDL number MFCD18447682
  • Cas Number : 1287752-87-7
  • Molecular Weight : 209.0251

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043453

Safety Information

MSDS https://www.hit2lead.com/sds/4043453_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart