CH4312834726 ChemBridge Corporation

1-isopropyl-1H-pyrrole-2-carbonitrile

  • MDL number MFCD18447683
  • Cas Number : 101001-62-1
  • Molecular Weight : 134.1784

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043479

Safety Information

MSDS https://www.hit2lead.com/sds/4043479_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart