CH4312834729 ChemBridge Corporation

2-methyl-5-(methylsulfonyl)aniline

  • MDL number MFCD09258802
  • Cas Number : 1671-48-3
  • Molecular Weight : 185.2434

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043521

Safety Information

MSDS https://www.hit2lead.com/sds/4043521_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart