CH4312834748 ChemBridge Corporation

4-chloro-3-methylisoxazolo[5,4-d]pyrimidine

  • MDL number MFCD14704902
  • Cas Number : 51850-60-3
  • Molecular Weight : 169.5685

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043747

Safety Information

MSDS https://www.hit2lead.com/sds/4043747_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart