CH4312834752 ChemBridge Corporation

4-aminopyrazolo[5,1-c][1,2,4]triazine-3-carbonitrile

  • MDL number MFCD03829962
  • Cas Number : 58390-42-4
  • Molecular Weight : 160.1362

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043784

Safety Information

MSDS https://www.hit2lead.com/sds/4043784_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart