CH4312834767 ChemBridge Corporation

(2-aminophenyl)dimethylamine

  • MDL number MFCD01706706
  • Molecular Weight : 136.1943

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5657224

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/building-blocks/5657224
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart