CH4321848527 ChemBridge Corporation

tert-butyl rac-(1R,5S,6r)-6-formyl-3-azabicyclo[3.1.0]hexane-3-carboxylate

  • MDL number MFCD18782860
  • Cas Number : 419572-19-3
  • Molecular Weight : 211.2576

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042366

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4042366
Availability details for
Quantity
  • Availability
  • Add to Cart