CH4321848533 ChemBridge Corporation

1-cyclopropyl-1H-pyrrole-2-carbonitrile

  • MDL number MFCD18917025
  • Cas Number : 1308384-55-5
  • Molecular Weight : 132.1625

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042617

Safety Information

MSDS https://www.hit2lead.com/sds/4042617_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart