CH4321848542 ChemBridge Corporation

[3-(trifluoromethyl)-1,2,4-oxadiazol-5-yl]acetonitrile

  • MDL number MFCD18917038
  • Cas Number : 1308384-47-5
  • Molecular Weight : 177.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043533

Safety Information

MSDS https://www.hit2lead.com/sds/4043533_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart