CH4321848634 ChemBridge Corporation

4-(3-methoxyphenyl)piperidine

  • MDL number MFCD05189908
  • Cas Number : 99329-55-2
  • Molecular Weight : 191.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3002188

Safety Information

MSDS https://www.hit2lead.com/sds/3002188_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart