CH4321848635 ChemBridge Corporation

4-formylbenzenesulfonamide

  • MDL number MFCD03196508
  • Cas Number : 3240-35-5
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/3002297

Safety Information

MSDS https://www.hit2lead.com/sds/3002297_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart