CH4321848659 ChemBridge Corporation

5-amino-3-methyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD02020550
  • Molecular Weight : 122.1279

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042595

Safety Information

MSDS https://www.hit2lead.com/sds/4042595_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart