CH4321848660 ChemBridge Corporation

3-(4-pyridinyl)-1H-1,2,4-triazol-5-amine

  • MDL number MFCD00465601
  • Cas Number : 3652-17-3
  • Molecular Weight : 161.164

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014290

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4014290
Availability details for
Quantity
  • Availability
  • Add to Cart