CH4321848680 ChemBridge Corporation

5-(3,4-dimethyl-5-isoxazolyl)-2-thiophenesulfonyl chloride

  • MDL number MFCD15729905
  • Cas Number : 1279219-52-1
  • Molecular Weight : 277.7477

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042882

Safety Information

signalword Danger
hcodes 8
MSDS https://www.hit2lead.com/sds/4042882_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart