CH4321848686 ChemBridge Corporation

azacyclotridecane

  • MDL number MFCD00022324
  • Cas Number : 295-03-4
  • Molecular Weight : 183.3336

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042914

Safety Information

MSDS https://www.hit2lead.com/sds/4042914_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart