CH4321848690 ChemBridge Corporation

3H-imidazo[4,5-b]pyridine-2-carboxylic acid

  • MDL number MFCD05721719
  • Cas Number : 97640-15-8
  • Molecular Weight : 163.1335

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042970

Safety Information

MSDS https://www.hit2lead.com/sds/4042970_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart