CH4321848703 ChemBridge Corporation

3-isopropyl-4-methoxybenzenesulfonyl chloride

  • MDL number MFCD11133929
  • Cas Number : 627082-12-6
  • Molecular Weight : 248.7264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043074

Safety Information

signalword Danger
hcodes 8
MSDS https://www.hit2lead.com/sds/4043074_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart