CH4321848708 ChemBridge Corporation

1-(1-methylcyclohexyl)methanamine

  • MDL number MFCD09607978
  • Cas Number : 3913-98-2
  • Molecular Weight : 127.2273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043142

Safety Information

MSDS https://www.hit2lead.com/sds/4043142_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart