CH4321848710 ChemBridge Corporation

1-cyclopropyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD12181391
  • Cas Number : 1155519-52-0
  • Molecular Weight : 151.1626

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4043147

Safety Information

MSDS https://www.hit2lead.com/sds/4043147_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart